siRNA Details
Gene Symbol: Smad6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008542.3 | NP_032568.3 | 4 | 2194 | ||
GenBank mRNA | AB221689.1 | - | 4 | 1159 | ||
AF010133.1 | - | 4 | 1159 | |||
AK004671.1 | - | 4 | 640 | |||
AK046602.1 | - | 4 | 2197 | |||
BC047280.1 | - | 4 | 1456 | 6477199 |
