siRNA Details
Gene Symbol: Mag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010758.2 | NP_034888.1 | 8 | 1501 | ||
GenBank mRNA | BC101944.1 | - | 8 | 1474 | 40039200 | |
BC103521.1 | - | 8 | 1474 | 40039204 | ||
BC103522.1 | - | 8 | 1474 | |||
BC103523.1 | - | 9 | 1348 | |||
M31811.1 | - | 8 | 1501 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
