siRNA Details
Gene Symbol: Mapkapk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010765.2 | NP_034895.1 | 13, 14 | 2027 | ||
GenBank mRNA | AF039840.1 | - | 13, 14 | 2041 | ||
AY533679.1 | - | 13, 14 | 1329 | |||
AY533681.1 | - | 8, 9 | 888 | |||
AY533682.1 | - | 8, 9 | 882 | |||
BC019184.1 | - | 13, 14 | 1449 | 5010305 |
