siRNA Details
Gene Symbol: Ascl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008553.4 | NP_032579.2 | 1 | 1158 | ||
GenBank mRNA | AK141866.1 | - | 1 | 1148 | ||
AK143210.1 | - | 1 | 1159 | |||
BC055748.1 | - | 1 | 680 | 6415061 | ||
JX478273.1 | - | 1 | 586 | |||
M95603.1 | - | 1 | 661 |

More Information
Other Species Targeted

Gene Ontology Categories:
