siRNA Details
Gene Symbol: Masp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003893.2 | NP_001003893.1 | 2 | 161 | ||
NM_010767.3 | NP_034897.1 | 2 | 161 | |||
GenBank mRNA | AA238260.1 | - | 2 | 149 | ||
AB009459.1 | - | 2 | 151 | |||
AI529228.1 | - | 2 | 162 | |||
AJ250369.1 | - | 2 | 150 | |||
AK050052.1 | - | 2 | 138 | |||
AK149421.1 | - | 1 | 215 | |||
BC013893.1 | - | 1 | 135 | 4211293 | ||
BF233497.1 | - | 2 | 159 | |||
Y19160.1 | - | 1 | 208 | |||
Y19163.1 | - | 1 | 178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
