siRNA Details
Gene Symbol: Max | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146176.1 | NP_001139648.1 | 2 | 287 | ||
NM_008558.2 | NP_032584.2 | 3 | 314 | |||
GenBank mRNA | AK080431.1 | - | 2 | 242 | ||
BC138671.1 | - | 3 | 170 | 8861692 | ||
BC138672.1 | - | 3 | 170 | 8861693 | ||
BC145369.1 | - | 2 | 143 | 9053927 | ||
M63903.1 | - | 3 | 251 |

More Information
Other Species Targeted

Gene Ontology Categories:
