siRNA Details
Gene Symbol: Mb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164047.1 | NP_001157519.1 | 2 | 326 | ||
NM_001164048.1 | NP_001157520.1 | 3 | 186 | |||
NM_013593.3 | NP_038621.2 | 3 | 274 | |||
GenBank mRNA | AA271284.1 | - | 3 | 186 | ||
AK137456.1 | - | 2 | 181 | |||
BC025172.1 | - | 3 | 286 | 3979003 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
