siRNA Details
Gene Symbol: Mc2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271716.1 | NP_001258645.1 | 4 | 825 | ||
NM_001271717.1 | NP_001258646.1 | 3 | 737 | |||
NM_001301372.1 | NP_001288301.1 | 5 | 911 | |||
NM_008560.3 | NP_032586.1 | 3 | 768 | |||
GenBank mRNA | AK033100.1 | - | 3 | 707 | ||
BC139210.1 | - | 2 | 671 | 8862232 | ||
BC139211.1 | - | 2 | 670 | 8862233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
