siRNA Details
Gene Symbol: Mcf2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159485.1 | NP_001152957.1 | 12 | 1646 | ||
NM_001159486.1 | NP_001152958.1 | 12 | 1646 | |||
NM_178076.3 | NP_835177.2 | 12 | 1549 | |||
GenBank mRNA | AK122259.1 | - | 3 | 454 | ||
AK147252.1 | - | 9 | 1753 | |||
AK170186.1 | - | 12 | 1607 | |||
AK170558.1 | - | 12 | 1580 | |||
BC058622.1 | - | 12 | 1484 | 5704822 | ||
DQ978380.1 | - | 11 | 1400 | |||
S76838.1 | - | 13 | 1843 |
