siRNA Details
Gene Symbol: Cd46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010778.3 | NP_034908.1 | 3, 4 | 445 | ||
GenBank mRNA | AB001566.1 | - | 3, 4 | 422 | ||
AB010919.1 | - | 3, 4 | 442 | |||
AB022600.1 | - | 3, 4 | 422 | |||
AK006632.1 | - | 3, 4 | 446 | |||
BC150660.1 | - | 3, 4 | 446 | 9087571 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
