siRNA Details
Gene Symbol: Cd46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010778.3 | NP_034908.1 | 2 | 225 | ||
GenBank mRNA | AB001566.1 | - | 2 | 202 | ||
AB010919.1 | - | 2 | 222 | |||
AB022600.1 | - | 2 | 202 | |||
AK006632.1 | - | 2 | 226 | |||
BC150660.1 | - | 2 | 226 | 9087571 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
