siRNA Details
Gene Symbol: Mcpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008570.1 | NP_032596.1 | 3 | 306 | ||
GenBank mRNA | BC100700.1 | - | 3 | 297 | 40043851 | |
BC100701.1 | - | 3 | 297 | 40043850 | ||
BC100702.1 | - | 3 | 297 | 40043849 | ||
BC100703.1 | - | 3 | 297 | 40043848 | ||
S44609.1 | - | 3 | 299 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
