siRNA Details
Gene Symbol: Mcpt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010779.2 | NP_034909.2 | 3 | 325 | ||
GenBank mRNA | AB051900.1 | - | 3 | 273 | ||
AK144385.1 | - | 3 | 326 | |||
AY007569.1 | - | 3 | 325 | |||
BC026198.1 | - | 3 | 301 | 1314753 | ||
CJ046459.1 | - | 3 | 328 | |||
M55617.1 | - | 325 | ||||
X68804.1 | - | 3 | 272 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
