siRNA Details
Gene Symbol: Mgrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252437.1 | NP_001239366.1 | 3, 4 | 593 | ||
NM_029657.4 | NP_083933.1 | 3, 4 | 590 | |||
GenBank mRNA | AK011747.2 | - | 3, 4 | 383 | ||
AK088533.1 | - | 3, 4 | 373 | |||
AK150176.1 | - | 4, 5 | 426 | |||
AK153407.1 | - | 3, 4 | 414 | |||
BC046830.1 | - | 3, 4 | 439 | 6811606 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
