siRNA Details
Gene Symbol: Meis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193271.1 | NP_001180200.1 | 6 | 1288 | ||
NM_010789.3 | NP_034919.1 | 6 | 1288 | |||
GenBank mRNA | AB221660.1 | - | 5 | 551 | ||
AK032947.1 | - | 5 | 1099 | |||
AK049720.1 | - | 5 | 1100 | |||
AK132298.1 | - | 6 | 1068 | |||
AK134321.1 | - | 6 | 1283 | |||
AK140748.1 | - | 6 | 1292 | |||
AK146680.1 | - | 2 | 96 | |||
AK168684.1 | - | 6 | 1280 | |||
BC023689.1 | - | 6 | 1284 | 5344307 | ||
U33629.1 | - | 6 | 999 | |||
U33630.1 | - | 6 | 999 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001134702, XM_006251529, BC161984.
Gene Ontology Categories:
