siRNA Details
Gene Symbol: Meox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008584.3 | NP_032610.1 | 2 | 935 | ||
GenBank mRNA | AK028352.1 | - | 2 | 936 | ||
AK031152.1 | - | 2 | 892 | |||
AK142690.1 | - | 2 | 814 | |||
BC002076.1 | - | 2 | 834 | 3591924 | ||
Z16406.1 | - | 2 | 813 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
