siRNA Details
Gene Symbol: Mesp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008589.2 | NP_032615.2 | 1 | 800 | ||
GenBank mRNA | AK146740.1 | - | 1 | 280 | ||
BC145652.1 | - | 1 | 630 | 40130955 | ||
BC145654.1 | - | 1 | 630 | 40130957 | ||
U71125.1 | - | 1 | 805 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
