siRNA Details
Gene Symbol: Mest | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252292.1 | NP_001239221.1 | 6 | 815 | ||
NM_001252293.1 | NP_001239222.1 | 5 | 536 | |||
NM_008590.2 | NP_032616.1 | 5 | 714 | |||
GenBank mRNA | AF482999.1 | - | 6 | 536 | ||
AK031718.1 | - | 5 | 713 | |||
AK032881.1 | - | 5 | 713 | |||
AK034949.1 | - | 5 | 712 | |||
AK146457.1 | - | 6 | 815 | |||
AK168714.1 | - | 5 | 712 | |||
AK168732.1 | - | 5 | 712 | |||
AK168743.1 | - | 5 | 712 | |||
AK168995.1 | - | 5 | 712 | |||
AK169095.1 | - | 5 | 712 | |||
AK169266.1 | - | 5 | 712 | |||
BC006639.1 | - | 5 | 697 | 3482928 | ||
CF916557.1 | - | 5 | 528 | |||
D16262.1 | - | 5 | 701 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
