siRNA Details
Gene Symbol: Foxc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008592.2 | NP_032618.2 | 1 | 1978 | ||
GenBank mRNA | AF045017.1 | - | 1 | 1965 | ||
AK144420.1 | - | 1 | 1881 | |||
BC052011.1 | - | 1 | 1816 | 5720391 |

More Information
Other Species Targeted

Gene Ontology Categories:
