siRNA Details
Gene Symbol: Mfge8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045489.1 | NP_001038954.1 | 6 | 787 | ||
NM_008594.2 | NP_032620.2 | 7 | 898 | |||
GenBank mRNA | AB021130.2 | - | 7 | 805 | ||
AB025280.1 | - | 6 | 694 | |||
AK087555.1 | - | 6 | 777 | |||
AK089211.1 | - | 6 | 753 | |||
AK145183.1 | - | 6 | 751 | |||
AK148371.1 | - | 6 | 776 | |||
AK152088.1 | - | 7 | 887 | |||
AK155047.1 | - | 7 | 896 | |||
AK169997.1 | - | 6 | 751 | |||
AK170935.1 | - | 6 | 751 | |||
AK171143.1 | - | 7 | 862 | |||
AK189618.1 | - | 2 | 172 | |||
AK207561.1 | - | 1 | 44 | |||
AK212128.1 | - | 1 | 44 | |||
AK212659.1 | - | 1 | 44 | |||
AK213407.1 | - | 1 | 44 | |||
AK214404.1 | - | 1 | 44 | |||
AK219710.1 | - | 1 | 44 | |||
BC003892.1 | - | 6 | 731 | 3593310 | ||
BC003904.1 | - | 6 | 726 | 3601565 | ||
BC018577.1 | - | 6 | 724 | |||
BC037475.1 | - | 3 | 1113 | |||
M38337.1 | - | 7 | 850 | |||
Y11684.2 | - | 6 | 694 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
