siRNA Details
Gene Symbol: Mgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110148.1 | NP_001103618.1 | 4 | 1766 | ||
NM_001110149.1 | NP_001103619.1 | 2 | 1122 | |||
NM_001110150.1 | NP_001103620.1 | 3 | 1296 | |||
NM_010794.3 | NP_034924.3 | 3 | 1641 | |||
GenBank mRNA | AK004760.1 | - | 3 | 1296 | ||
AK040121.1 | - | 2 | 1123 | |||
AK087959.1 | - | 3 | 1260 | |||
AK155322.1 | - | 3 | 1337 | |||
AK179728.1 | - | 1 | 49 | |||
BC006629.1 | - | 3 | 1245 | 3969155 | ||
BC031752.1 | - | 3 | 1586 | 4483519 | ||
L07037.1 | - | 3 | 1309 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
