siRNA Details
Gene Symbol: Minpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010799.2 | NP_034929.1 | 2 | 884 | ||
GenBank mRNA | AB041574.1 | - | 2 | 717 | ||
AF046908.1 | - | 2 | 884 | |||
AK042453.1 | - | 2 | 731 | |||
AK128911.1 | - | 2 | 763 | |||
AK151486.1 | - | 2 | 729 | |||
AK181609.1 | - | 1 | 101 | |||
AK183676.1 | - | 2 | 144 | |||
BC021437.1 | - | 2 | 785 | 5039704 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
