siRNA Details
Gene Symbol: Minpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010799.2 | NP_034929.1 | 5 | 1371 | ||
GenBank mRNA | AB041574.1 | - | 5 | 1204 | ||
AF046908.1 | - | 5 | 1371 | |||
AK128911.1 | - | 5 | 1250 | |||
AK151486.1 | - | 5 | 1216 | |||
BC021437.1 | - | 5 | 1272 | 5039704 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
