siRNA Details
Gene Symbol: Mitf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113198.1 | NP_001106669.1 | 7, 8 | 1073 | ||
NM_001178049.1 | NP_001171520.1 | 7, 8 | 960 | |||
NM_008601.3 | NP_032627.1 | 6, 7 | 764 | |||
GenBank mRNA | AB221657.1 | - | 6, 7 | 626 | ||
AK052895.1 | - | 7, 8 | 932 | |||
AK154189.1 | - | 7, 8 | 1074 | |||
AK155350.1 | - | 7, 8 | 1074 | |||
BC108976.2 | - | 6, 7 | 728 | 40047440 | ||
BC108977.1 | - | 6, 7 | 693 | 40047441 | ||
Z23066.1 | - | 6, 7 | 755 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
