siRNA Details
Gene Symbol: Trpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039104.2 | NP_001034193.2 | 25 | 3620 | ||
GenBank mRNA | AJ867481.1 | - | 6 | 875 | ||
AK148193.1 | - | 23 | 3462 | |||
AY180104.2 | - | 25 | 3620 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
