siRNA Details
Gene Symbol: Ndst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010811.2 | NP_034941.2 | 8 | 2457 | ||
GenBank mRNA | AF074925.1 | - | 6 | 1707 | ||
AK150100.1 | - | 8 | 2238 | |||
BC050829.1 | - | 8 | 2222 | |||
BC110479.1 | - | 6 | 1793 | 40048114 | ||
BC110480.1 | - | 6 | 1855 | 40048115 | ||
U02304.1 | - | 7 | 2219 | |||
X75885.1 | - | 6 | 1861 |
