siRNA Details
Gene Symbol: Mobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039364.2 | NP_001034453.1 | 2 | 208 | ||
NM_001039365.2 | NP_001034454.1 | 3 | 287 | |||
NM_008614.2 | NP_032640.1 | 3 | 287 | |||
GenBank mRNA | AK013799.1 | - | 3 | 288 | ||
AK019323.1 | - | 3 | 288 | |||
AK028057.1 | - | 3 | 287 | |||
AK083103.1 | - | 3 | 288 | |||
AK140055.1 | - | 3 | 288 | |||
BC048492.1 | - | 3 | 287 | 6593408 | ||
BY257495.1 | - | 2 | 209 | |||
U81317.1 | - | 2 | 207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
