siRNA Details
Gene Symbol: Mocs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113374.1 | NP_001106845.1 | 4 | 408 | ||
NM_013826.3 | NP_038854.2 | 5 | 530 | |||
GenBank mRNA | AF091872.1 | - | 5 | 481 | ||
AK002719.1 | - | 4 | 376 | |||
AK010112.1 | - | 5 | 510 | |||
AK078782.1 | - | 4 | 376 | |||
BC004631.1 | - | 4 | 379 | |||
BC024371.1 | - | 5 | 490 | 4983231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
