siRNA Details
Gene Symbol: Msx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010835.2 | NP_034965.2 | 2 | 1393 | ||
GenBank mRNA | AK077524.1 | - | 2 | 1396 | ||
AK078600.1 | - | 2 | 1394 | |||
AK145726.1 | - | 2 | 1400 | |||
BC016426.1 | - | 2 | 1376 | 4923403 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
