siRNA Details
Gene Symbol: Grpel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024478.2 | NP_077798.1 | 4 | 379 | ||
GenBank mRNA | AK048732.1 | - | 4 | 379 | ||
AK050445.1 | - | 4 | 374 | |||
AK089113.1 | - | 4 | 375 | |||
AK139561.1 | - | 4 | 567 | |||
AK161688.1 | - | 4 | 392 | |||
BC002284.1 | - | 4 | 429 | 3589725 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
