siRNA Details
Gene Symbol: Grpel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021296.2 | NP_067271.1 | 4 | 494 | ||
GenBank mRNA | AF041060.1 | - | 4 | 459 | ||
AK003011.1 | - | 4 | 475 | |||
AK012634.1 | - | 4 | 473 | |||
AK082983.1 | - | 484 | ||||
AK134455.1 | - | 4 | 473 | |||
AK145493.1 | - | 4 | 486 | |||
BC068232.1 | - | 4 | 471 | 30533079 | ||
BC094583.1 | - | 4 | 461 | |||
BC120747.1 | - | 4 | 484 | 40129670 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
