siRNA Details
Gene Symbol: Map1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008634.2 | NP_032660.2 | 5 | 7093 | ||
GenBank mRNA | AK148870.1 | - | 1 | 558 | ||
BC138033.1 | - | 5 | 7023 | 8861052 | ||
BC138034.1 | - | 5 | 7023 | 8861053 | ||
X51396.1 | - | 5 | 7031 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
