siRNA Details
Gene Symbol: Mtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008636.4 | NP_032662.3 | 2 | 294 | ||
GenBank mRNA | AK012676.1 | - | 2 | 262 | ||
AK030284.1 | - | 2 | 273 | |||
AK150295.1 | - | 3 | 356 | |||
AK208188.1 | - | 1 | 156 | |||
AK211875.1 | - | 1 | 156 | |||
AK213643.1 | - | 1 | 156 | |||
AY260849.1 | - | 1 | 148 | |||
AY260850.1 | - | 1 | 148 | |||
BC017679.1 | - | 2 | 256 | 4210829 | ||
DV658365.1 | - | 2 | 313 | |||
X71327.1 | - | 2 | 244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
