siRNA Details
Gene Symbol: Mtm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164190.1 | NP_001157662.1 | 11 | 1386 | ||
NM_001164191.1 | NP_001157663.1 | 11 | 1277 | |||
NM_001164192.1 | NP_001157664.1 | 11 | 1277 | |||
NM_019926.3 | NP_064310.2 | 11 | 1266 | |||
GenBank mRNA | AF073996.1 | - | 11 | 1267 | ||
AK016445.1 | - | 11 | 1387 | |||
AK166452.1 | - | 11 | 1245 | |||
AK171475.1 | - | 11 | 1256 | |||
AK220524.1 | - | 3387 | ||||
BC051022.1 | - | 11 | 1235 | |||
BC066847.1 | - | 11 | 1224 | |||
BC090984.1 | - | 11 | 1244 | 30246206 |
