siRNA Details
Gene Symbol: Mtnr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008639.2 | NP_032665.1 | 2 | 580 | ||
GenBank mRNA | BC116900.1 | - | 2 | 653 | 40126219 | |
BC119313.1 | - | 2 | 653 | 8734062 | ||
U52222.1 | - | 2 | 580 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
