siRNA Details
Gene Symbol: Mup15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200004.1 | NP_001186933.1 | 7 | 759 |
Gene Symbol: Gm2083 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134644.1 | NP_001128116.1 | 7 | 752 |
Gene Symbol: Mup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163011.1 | NP_001156483.1 | 7 | 839 | ||
NM_031188.2 | NP_112465.2 | 7 | 798 | |||
GenBank mRNA | BC012221.1 | - | 7 | 744 | 4195753 | |
BC013649.1 | - | 732 | 4195401 | |||
BC091744.1 | - | 735 | 5102271 | |||
BC091775.1 | - | 7 | 745 | 5137341 | ||
BC092254.1 | - | 732 | 5100458 | |||
BC093515.1 | - | 2030 | ||||
BC099597.1 | - | 735 | 947916 | |||
BC100586.1 | - | 752 | 6769126 | |||
BC106092.1 | - | 7 | 735 | 5098340 | ||
M16355.1 | - | 7 | 798 | |||
M27609.1 | - | 577 | ||||
M28649.1 | - | 745 | ||||
X03524.1 | - | 600 | ||||
X04115.1 | - | 556 |
Gene Symbol: Mup10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122647.1 | NP_001116119.1 | 7 | 754 |
Gene Symbol: Mup12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199995.1 | NP_001186924.1 | 7 | 753 |
Gene Symbol: Mup13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134674.1 | NP_001128146.1 | 7 | 854 |
Gene Symbol: Mup14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199999.1 | NP_001186928.1 | 7 | 796 |
Gene Symbol: Mup19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135127.2 | NP_001128599.1 | 7 | 753 | ||
GenBank mRNA | AK013259.1 | - | 7 | 753 |
Gene Symbol: Mup2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286096.1 | NP_001273025.1 | 6 | 623 | ||
NM_008647.4 | NP_032673.3 | 7 | 757 | |||
GenBank mRNA | BC012259.1 | - | 746 | 4196403 | ||
BC059097.1 | - | 732 | 4159984 | |||
M16356.1 | - | 745 |
Gene Symbol: Mup7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134675.1 | NP_001128147.1 | 7 | 903 |
Gene Symbol: Mup8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134676.1 | NP_001128148.1 | 7 | 858 |
Gene Symbol: Mup9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281979.1 | NP_001268908.1 | 7 | 753 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC094452.1 | - | 733 | Chr. - - [-] on Build |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
