siRNA Details
Gene Symbol: Mup3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039544.1 | NP_001034633.1 | 3 | 359 | ||
GenBank mRNA | AI046324.1 | - | 3 | 417 | ||
BC019965.1 | - | 3 | 321 | 4194688 | ||
BC059102.1 | - | 3 | 336 | 4195272 | ||
BC106100.1 | - | 3 | 325 | 5099753 | ||
EU882235.1 | - | 3 | 278 | |||
M27608.1 | - | 3 | 308 | |||
X03525.1 | - | 3 | 307 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
