siRNA Details
Gene Symbol: Myh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177369.3 | NP_796343.2 | 3 | 368 | ||
GenBank mRNA | AK029328.1 | - | 3 | 279 | ||
AK081482.1 | - | 3 | 279 | |||
AK087387.1 | - | 3 | 279 | |||
AY963802.1 | - | 3 | 278 | |||
BC150737.1 | - | 3 | 291 | 9087648 | ||
DQ021876.1 | - | 3 | 318 | |||
DQ021877.1 | - | 3 | 327 | |||
DQ021878.1 | - | 2 | 321 | |||
DQ021879.1 | - | 1 | 256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
