siRNA Details
Gene Symbol: Myh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177369.3 | NP_796343.2 | 20 | 2439 | ||
GenBank mRNA | AK029328.1 | - | 20 | 2350 | ||
BC150737.1 | - | 20 | 2362 | 9087648 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
