siRNA Details
Gene Symbol: Myod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010866.2 | NP_034996.2 | 3 | 1772 | ||
GenBank mRNA | AK076157.1 | - | 3 | 1773 | ||
AK142859.1 | - | 3 | 1774 | |||
M18779.1 | - | 3 | 1732 | |||
M84918.1 | - | 3 | 1764 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
