siRNA Details
Gene Symbol: Myom2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008664.2 | NP_032690.2 | 12 | 1473 | ||
GenBank mRNA | AJ001038.1 | - | 12 | 1473 | ||
AK142174.1 | - | 12 | 1449 | |||
BC115722.1 | - | 12 | 1464 | 40086642 | ||
BC115834.1 | - | 12 | 1463 | 40086646 |

More Information
Other Species Targeted

Gene Ontology Categories:
