siRNA Details
Gene Symbol: Myom2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008664.2 | NP_032690.2 | 29 | 3601 | ||
GenBank mRNA | AJ001038.1 | - | 29 | 3601 | ||
AK142174.1 | - | 29 | 3577 | |||
BC115722.1 | - | 29 | 3592 | 40086642 | ||
BC115834.1 | - | 29 | 3591 | 40086646 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
