siRNA Details
Gene Symbol: Naca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113199.1 | NP_001106670.1 | 8 | 6782 | ||
NM_001282976.1 | NP_001269905.1 | 7 | 977 | |||
NM_013608.3 | NP_038636.2 | 7 | 866 | |||
GenBank mRNA | AK146149.1 | - | 7 | 570 | ||
AK146197.1 | - | 7 | 604 | |||
AK151928.1 | - | 7 | 568 | |||
AK161678.1 | - | 7 | 561 | |||
AK168023.1 | - | 7 | 579 | |||
AK168169.1 | - | 7 | 568 | |||
AK168607.1 | - | 7 | 548 | |||
AK169432.1 | - | 7 | 638 | |||
AK169435.1 | - | 7 | 568 | |||
BC029830.1 | - | 7 | 556 | |||
BC083340.1 | - | 7 | 551 | 5700166 | ||
BC099375.1 | - | 6 | 541 | 30525972 | ||
BI851172.1 | - | 4 | 353 | |||
U22151.1 | - | 6 | 542 | |||
U48364.1 | - | 8 | 6493 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
