siRNA Details
Gene Symbol: Naca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282976.1 | NP_001269905.1 | 2, 3 | 520 | ||
NM_013608.3 | NP_038636.2 | 2, 3 | 409 | |||
GenBank mRNA | AK146149.1 | - | 2, 3 | 113 | ||
AK146197.1 | - | 2, 3 | 147 | |||
AK151928.1 | - | 2, 3 | 111 | |||
AK161678.1 | - | 2, 3 | 104 | |||
AK168023.1 | - | 2, 3 | 122 | |||
AK168169.1 | - | 2, 3 | 111 | |||
AK168607.1 | - | 2, 3 | 91 | |||
AK169432.1 | - | 2, 3 | 181 | |||
AK169435.1 | - | 2, 3 | 111 | |||
AL355707.1 | - | 2, 3 | 142 | |||
BC029830.1 | - | 2, 3 | 99 | |||
BC083340.1 | - | 2, 3 | 94 | 5700166 | ||
BC099375.1 | - | 1, 2 | 84 | 30525972 | ||
BU962928.1 | - | 2, 3 | 383 | |||
CN840834.1 | - | 412 | ||||
CX236470.1 | - | 2, 3 | 245 | |||
U22151.1 | - | 1, 2 | 85 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
