siRNA Details
Gene Symbol: Nap1l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285489.1 | NP_001272418.1 | 7 | 582 | ||
NM_001285490.1 | NP_001272419.1 | 7 | 595 | |||
NM_008672.3 | NP_032698.1 | 7 | 582 | |||
GenBank mRNA | AJ002198.1 | - | 7 | 552 | ||
AK090112.1 | - | 7 | 598 | |||
AK219914.1 | - | 2 | 148 | |||
BC089018.1 | - | 7 | 568 | |||
BC132039.1 | - | 6 | 506 | 40130316 | ||
BC138518.1 | - | 6 | 506 | 8861538 | ||
BC145306.1 | - | 6 | 506 | 9053860 | ||
BC145308.1 | - | 6 | 506 | 9053862 |
