siRNA Details
Gene Symbol: Ndrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008681.2 | NP_032707.2 | 8, 9 | 689 | ||
GenBank mRNA | AK004548.1 | - | 8, 9 | 689 | ||
AK098075.1 | - | 8, 9 | 688 | |||
AK161860.1 | - | 8, 9 | 691 | |||
AK165007.1 | - | 8, 9 | 689 | |||
AK170281.1 | - | 8, 9 | 690 | |||
AK170439.1 | - | 8, 9 | 688 | |||
AK170636.1 | - | 8, 9 | 688 | |||
AK170809.1 | - | 8, 9 | 689 | |||
BC015282.1 | - | 8, 9 | 644 | 4237943 | ||
BC071235.1 | - | 8, 9 | 697 | 4242075 | ||
BC141246.1 | - | 8, 9 | 580 | 9055858 | ||
BC171968.1 | - | 8, 9 | 580 | 9054254 | ||
CA320478.1 | - | 5, 6 | 432 | |||
U52073.1 | - | 8, 9 | 665 | |||
U60593.1 | - | 8, 9 | 687 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
