siRNA Details
Gene Symbol: Nek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF007247.1 | - | 2, 3 | 404 | ||
AF013166.1 | - | 2, 3 | 374 | |||
AK170179.1 | - | 2, 3 | 410 | |||
AK171277.1 | - | 2, 3 | 410 | |||
AK171387.1 | - | 2, 3 | 410 | |||
BC010302.1 | - | 2, 3 | 412 | 3597227 | ||
BC011316.1 | - | 2, 3 | 410 | 3674001 | ||
U95610.1 | - | 2, 3 | 401 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
