siRNA Details
Gene Symbol: Nek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010892.3 | NP_035022.2 | 3 | 685 | ||
GenBank mRNA | AF007247.1 | - | 3 | 599 | ||
AF013166.1 | - | 3 | 569 | |||
AK077627.1 | - | 3 | 649 | |||
AK147072.1 | - | 3 | 661 | |||
AK164467.1 | - | 3 | 628 | |||
AK169330.1 | - | 4 | 576 | |||
AK170179.1 | - | 3 | 605 | |||
AK171277.1 | - | 3 | 605 | |||
AK171387.1 | - | 3 | 605 | |||
BC010302.1 | - | 3 | 607 | 3597227 | ||
BC011316.1 | - | 3 | 605 | 3674001 | ||
BC057576.1 | - | 3 | 625 | 6810914 | ||
U95610.1 | - | 3 | 596 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
