siRNA Details
Gene Symbol: Neo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042752.1 | NP_001036217.1 | 3 | 1049 | ||
NM_008684.2 | NP_032710.2 | 3 | 1049 | |||
GenBank mRNA | BC054540.1 | - | 3 | 1033 | 6406007 | |
Y09535.1 | - | 3 | 916 |
