siRNA Details
Gene Symbol: Neo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042752.1 | NP_001036217.1 | 17, 18 | 3068 | ||
NM_008684.2 | NP_032710.2 | 17, 18 | 3116 | |||
GenBank mRNA | BC054540.1 | - | 17, 18 | 3052 | 6406007 | |
CF539503.1 | - | 2, 3 | 268 | |||
CN457663.1 | - | 1, 2 | 85 | |||
Y09535.1 | - | 17, 18 | 2983 |
